Assay Details
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127544.1 | NP_001121016.1 | 7 - 8 | 987 | 77 | |
XM_008761404.1 | XP_008759626.1 | 6 - 7 | 1015 | 77 | ||
GenBank mRNA | BC162022.1 | - | 7 - 8 | 966 | 77 | |
FQ211892.1 | - | 6 - 7 | 947 | 77 | ||
FQ221591.1 | - | 7 - 8 | 1067 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
