Assay Details
Gene Symbol: Ap1ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191850.1 | NP_001178779.1 | 5 - 6 | 280 | 90 | |
XM_006233291.2 | XP_006233353.2 | 5 - 6 | 613 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
