Assay Details
Gene Symbol: Ap1ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191850.1 | NP_001178779.1 | 8 - 9 | 509 | 108 | |
XM_006233291.2 | XP_006233353.2 | 8 - 9 | 842 | 108 | ||
XM_006233292.2 | XP_006233354.2 | 7 - 8 | 743 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
