Assay Details
Gene Symbol: Bcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031328.1 | NP_112618.1 | 1 - 2 | 180 | 68 | |
GenBank mRNA | AB016069.1 | - | 1 - 2 | 180 | 68 | |
BC061772.1 | - | 1 - 2 | 168 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
