Assay Details
Gene Symbol: Frrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761474.1 | XP_008759696.1 | 3 - 4 | 236 | 65 | |
GenBank mRNA | FQ210266.1 | - | 3 - 4 | 173 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008761474.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
