Assay Details
Gene Symbol: Frrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761474.1 | XP_008759696.1 | 4 - 5 | 450 | 122 | |
GenBank mRNA | FQ210266.1 | - | 4 - 5 | 387 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
