Assay Details
Gene Symbol: Lamtor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008375.1 | NP_001008376.1 | 1 - 2 | 76 | 96 | |
GenBank mRNA | BC086353.1 | - | 1 - 2 | 76 | 96 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC086353.1, NM_001008375.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
