Assay Details
Gene Symbol: Lamtor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008375.1 | NP_001008376.1 | 3 - 4 | 165 | 74 | |
XM_006233383.2 | XP_006233445.1 | 3 - 4 | 290 | 74 | ||
XM_006233385.2 | XP_006233447.1 | 3 - 4 | 235 | 74 | ||
GenBank mRNA | BC086353.1 | - | 3 - 4 | 165 | 7304027 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
