Assay Details
Gene Symbol: Lamtor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008375.1 | NP_001008376.1 | 5 - 6 | 362 | 90 | |
XM_006233383.2 | XP_006233445.1 | 5 - 6 | 487 | 90 | ||
XM_006233385.2 | XP_006233447.1 | 5 - 6 | 432 | 90 | ||
GenBank mRNA | BC086353.1 | - | 5 - 6 | 362 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
