Assay Details
Gene Symbol: Dennd2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107714.1 | NP_001101184.1 | 2 - 3 | 321 | 107 | |
XM_006233114.2 | XP_006233176.1 | 2 - 3 | 270 | 107 | ||
XM_008761396.1 | XP_008759618.1 | 1 - 2 | 717 | 107 | ||
XM_008761397.1 | XP_008759619.1 | 1 - 2 | 717 | 107 | ||
XM_008761398.1 | XP_008759620.1 | 2 - 3 | 270 | 107 | ||
XM_008761399.1 | XP_008759621.1 | 2 - 3 | 354 | 107 | ||
GenBank mRNA | BC158880.1 | - | 2 - 3 | 257 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
