Assay Details
Gene Symbol: Fnbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039609.1 | NP_001034698.1 | 1505 | 80 | ||
GenBank mRNA | AB250295.1 | - | 1505 | 80 | ||
AB250296.1 | - | 1331 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
