Assay Details
Gene Symbol: Kat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750617.3 | XP_003750665.2 | 9 - 10 | 2458 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
