Assay Details
Gene Symbol: Kat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750617.3 | XP_003750665.2 | 3 - 4 | 1554 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
