Assay Details
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134459.1 | NP_604454.1 | 1 - 2 | 128 | 138 | |
XM_006251676.2 | XP_006251738.1 | 1 - 2 | 164 | 138 | ||
XM_008770596.1 | XP_008768818.1 | 1 - 2 | 154 | 138 | ||
GenBank mRNA | AB031014.1 | - | 1 - 2 | 128 | 138 | |
AF481858.1 | - | 1 - 2 | 75 | 138 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
