Assay Details
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251676.2 | XP_006251738.1 | 9 - 10 | 776 | 60 | |
XM_008770596.1 | XP_008768818.1 | 9 - 10 | 766 | 60 | ||
GenBank mRNA | AF481858.1 | - | 9 - 10 | 687 | 60 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
