Assay Details
Gene Symbol: Acaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130433.1 | NP_569117.1 | 8 - 9 | 1114 | 65 | |
GenBank mRNA | FQ216726.1 | - | 8 - 9 | 977 | 65 | |
FQ228855.1 | - | 8 - 9 | 978 | 65 | ||
X05341.1 | - | 8 - 9 | 1114 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
