Assay Details
Gene Symbol: Lipg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012741.1 | NP_001012759.1 | 4 - 5 | 577 | 116 | |
GenBank mRNA | AY916123.1 | - | 4 - 5 | 577 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
