Assay Details
Gene Symbol: Pcdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761025.1 | XP_008759247.1 | 4 - 5 | 3315 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
