Assay Details
Gene Symbol: Agpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133406.1 | NP_596897.1 | 7 - 8 | 1049 | 91 | |
XM_006227856.2 | XP_006227918.1 | 7 - 8 | 1764 | 91 | ||
XM_006227857.2 | XP_006227919.1 | 7 - 8 | 1418 | 91 | ||
GenBank mRNA | AB067572.1 | - | 7 - 8 | 1049 | 91 | |
BC086992.1 | - | 7 - 8 | 968 | 7191675 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
