Assay Details
Gene Symbol: Agpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133406.1 | NP_596897.1 | 8 - 9 | 1246 | 69 | |
XM_006227856.2 | XP_006227918.1 | 8 - 9 | 1961 | 69 | ||
XM_006227857.2 | XP_006227919.1 | 8 - 9 | 1615 | 69 | ||
GenBank mRNA | AB067572.1 | - | 8 - 9 | 1246 | 69 | |
BC086992.1 | - | 8 - 9 | 1165 | 7191675 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P13176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142689 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13176 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13176 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P13176 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13176 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68592 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103873 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2669 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74544 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7720 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28914 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66340 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10718006 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10718005 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10718009 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10718007 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10718008 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10718003 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10718004 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10718002 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10718001 | - | - |
Rat Genome 230 2.0 Array | 1368194_at | - | - |
Rat Genome 230 2.0 Array | 1368194_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368194_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368194_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100035478.rn.1 | TC0100004464.rn.1 | RTA TC |
Rat Expression Set 230 | 1368194_at | - | - |
Rat Expression Set 230 | 1368194_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10718001 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21397072 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372733 | - | - |
RatRef-12 Expression BeadChip | ILMN_56708 | - | - |
