Assay Details
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004078.1 | NP_001004078.1 | 1 - 2 | 110 | 81 | |
GenBank mRNA | BC059165.1 | - | 1 - 2 | 180 | 81 | |
BC079441.1 | - | 1 - 2 | 110 | 7115414 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
