Assay Details
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004078.1 | NP_001004078.1 | 4 - 5 | 531 | 88 | |
GenBank mRNA | AY683456.1 | - | 3 - 4 | 413 | 88 | |
BC059165.1 | - | 4 - 5 | 601 | 88 | ||
BC079441.1 | - | 4 - 5 | 531 | 7115414 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
