Assay Details
Gene Symbol: Cdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168631.1 | NP_001162102.1 | 5 - 6 | 1067 | 104 | |
XM_006232079.2 | XP_006232141.1 | 5 - 6 | 1143 | 104 | ||
XM_006232080.2 | XP_006232142.1 | 5 - 6 | 1175 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
