Assay Details
Gene Symbol: Epb41l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006227709.1 | XP_006227771.1 | 9 - 10 | 1570 | 126 | |
XM_008758646.1 | XP_008756868.1 | 9 - 10 | 1752 | 126 | ||
XM_008758647.1 | XP_008756869.1 | 9 - 10 | 1868 | 126 | ||
XM_008758648.1 | XP_008756870.1 | 9 - 10 | 1473 | 126 | ||
XM_008758649.1 | XP_008756871.1 | 10 - 11 | 1708 | 126 | ||
XM_008758650.1 | XP_008756872.1 | 9 - 10 | 4555 | 126 | ||
GenBank mRNA | FQ212032.1 | - | 10 - 11 | 1666 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
