Assay Details
Gene Symbol: Ets1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012555.2 | NP_036687.1 | 2 - 3 | 481 | 121 | |
XM_006242752.1 | XP_006242814.1 | 2 - 3 | 529 | 121 | ||
GenBank mRNA | BC101927.1 | - | 2 - 3 | 519 | 121 | |
L20681.1 | - | 2 - 3 | 496 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
