Assay Details
Gene Symbol: Snx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108131.1 | NP_001101601.1 | 2 - 3 | 1681 | 118 | |
XM_006242747.2 | XP_006242809.1 | 3 - 4 | 1998 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
