Assay Details
Gene Symbol: Snx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108131.1 | NP_001101601.1 | 6 - 7 | 2140 | 110 | |
XM_006242747.2 | XP_006242809.1 | 7 - 8 | 2457 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
