Assay Details
Gene Symbol: Npsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106808.1 | NP_001100278.1 | 5 - 6 | 568 | 72 | |
XM_008766049.1 | XP_008764271.1 | 4 - 5 | 811 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
