Assay Details
Gene Symbol: Npsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106808.1 | NP_001100278.1 | 7 - 8 | 844 | 59 | |
XM_008766049.1 | XP_008764271.1 | 6 - 7 | 1087 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
