Assay Details
Gene Symbol: Ntm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017354.1 | NP_059050.1 | 3 - 4 | 1009 | 74 | |
XM_006242749.1 | XP_006242811.1 | 3 - 4 | 1327 | 74 | ||
XM_008766065.1 | XP_008764287.1 | 3 - 4 | 1327 | 74 | ||
GenBank mRNA | U16845.1 | - | 3 - 4 | 1009 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
