Assay Details
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271227.1 | NP_001258156.1 | 1 - 2 | 398 | 145 | |
XM_006256722.1 | XP_006256784.1 | 1 - 2 | 393 | 145 | ||
XM_006256723.2 | XP_006256785.1 | 1 - 2 | 383 | 145 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006256723.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
