Assay Details
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271227.1 | NP_001258156.1 | 2 - 3 | 507 | 60 | |
XM_006256721.2 | XP_006256783.1 | 2 - 3 | 1152 | 60 | ||
XM_006256722.1 | XP_006256784.1 | 2 - 3 | 502 | 60 | ||
XM_006256727.2 | XP_006256789.1 | 2 - 3 | 516 | 60 | ||
XM_006256728.2 | XP_006256790.1 | 2 - 3 | 185 | 60 | ||
XM_006256729.2 | XP_006256791.1 | 2 - 3 | 136 | 60 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006256727.2,XM_006256728.2,XM_006256729.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
