Assay Details
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271227.1 | NP_001258156.1 | 3 - 4 | 810 | 56 | |
XM_006256721.2 | XP_006256783.1 | 3 - 4 | 1455 | 56 | ||
XM_006256722.1 | XP_006256784.1 | 3 - 4 | 805 | 56 | ||
XM_006256723.2 | XP_006256785.1 | 3 - 4 | 776 | 56 | ||
XM_006256725.2 | XP_006256787.1 | 3 - 4 | 469 | 56 | ||
XM_006256726.2 | XP_006256788.1 | 3 - 4 | 419 | 56 | ||
XM_006256727.2 | XP_006256789.1 | 3 - 4 | 819 | 56 | ||
XM_006256728.2 | XP_006256790.1 | 3 - 4 | 488 | 56 | ||
XM_006256729.2 | XP_006256791.1 | 3 - 4 | 439 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
