Assay Details
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271227.1 | NP_001258156.1 | 9 - 10 | 1561 | 157 | |
XM_006256721.2 | XP_006256783.1 | 9 - 10 | 2206 | 157 | ||
XM_006256722.1 | XP_006256784.1 | 8 - 9 | 1451 | 157 | ||
XM_006256723.2 | XP_006256785.1 | 9 - 10 | 1527 | 157 | ||
XM_006256725.2 | XP_006256787.1 | 9 - 10 | 1220 | 157 | ||
XM_006256726.2 | XP_006256788.1 | 9 - 10 | 1170 | 157 | ||
XM_006256727.2 | XP_006256789.1 | 9 - 10 | 1570 | 157 | ||
XM_006256728.2 | XP_006256790.1 | 9 - 10 | 1239 | 157 | ||
XM_006256729.2 | XP_006256791.1 | 9 - 10 | 1190 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
