Assay Details
Gene Symbol: Akap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024402.1 | NP_077378.1 | 1 - 2 | 86 | 63 | |
GenBank mRNA | AF008114.1 | - | 1 - 2 | 86 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF008114.1, NM_024402.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
