Assay Details
Gene Symbol: Npr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012868.1 | NP_037000.1 | 3 - 4 | 1059 | 85 | |
XM_006232045.2 | XP_006232107.1 | 3 - 4 | 1643 | 85 | ||
GenBank mRNA | L27339.1 | - | 3 - 4 | 1059 | 85 | |
X75605.1 | - | 3 - 4 | 1080 | 85 | ||
X78595.1 | - | 905 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
