Assay Details
Gene Symbol: Npr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012868.1 | NP_037000.1 | 1 - 2 | 768 | 165 | |
XM_006232045.2 | XP_006232107.1 | 1 - 2 | 1352 | 165 | ||
GenBank mRNA | L27339.1 | - | 1 - 2 | 768 | 165 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
