Assay Details
Gene Symbol: Prlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034111.1 | NP_001029283.1 | 7 - 8 | 862 | 112 | |
NM_012630.2 | NP_036762.1 | 7 - 8 | 872 | 112 | ||
XM_008760751.1 | XP_008758973.1 | 9 - 10 | 2469 | 112 | ||
GenBank mRNA | BC092133.1 | - | 7 - 8 | 866 | 112 | |
M19304.1 | - | 7 - 8 | 873 | 112 | ||
M57668.1 | - | 7 - 8 | 862 | 112 | ||
M74152.1 | - | 7 - 8 | 832 | 112 | ||
U07567.1 | - | 7 - 8 | 815 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
