Assay Details
Gene Symbol: Prlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034111.1 | NP_001029283.1 | 2 - 3 | 141 | 73 | |
GenBank mRNA | M57668.1 | - | 2 - 3 | 141 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the M57668.1, NM_001034111.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
