Assay Details
Gene Symbol: C1qtnf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232067.2 | XP_006232129.2 | 1 - 2 | 309 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
