Assay Details
Gene Symbol: Ppt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019367.2 | NP_062240.1 | 4 - 5 | 572 | 70 | |
XM_006256017.1 | XP_006256079.1 | 4 - 5 | 887 | 70 | ||
XM_006256018.2 | XP_006256080.1 | 4 - 5 | 1709 | 70 | ||
XM_006256019.2 | XP_006256081.1 | 4 - 5 | 1442 | 70 | ||
XM_006256020.2 | XP_006256082.1 | 4 - 5 | 1297 | 70 | ||
XM_006256022.2 | XP_006256084.1 | 4 - 5 | 1256 | 70 | ||
XM_006256023.2 | XP_006256085.1 | 4 - 5 | 730 | 70 | ||
XM_008772762.1 | XP_008770984.1 | 4 - 5 | 2196 | 70 | ||
GenBank mRNA | AF061971.1 | - | 4 - 5 | 504 | 70 | |
BC062023.1 | - | 4 - 5 | 534 | 70 | ||
FQ234132.1 | - | 4 - 5 | 514 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
