Assay Details
Gene Symbol: Ppt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019367.2 | NP_062240.1 | 6 - 7 | 766 | 131 | |
XM_006256017.1 | XP_006256079.1 | 6 - 7 | 1081 | 131 | ||
XM_006256018.2 | XP_006256080.1 | 6 - 7 | 1903 | 131 | ||
XM_006256019.2 | XP_006256081.1 | 6 - 7 | 1636 | 131 | ||
XM_006256020.2 | XP_006256082.1 | 6 - 7 | 1491 | 131 | ||
XM_006256022.2 | XP_006256084.1 | 6 - 7 | 1450 | 131 | ||
XM_006256023.2 | XP_006256085.1 | 6 - 7 | 924 | 131 | ||
XM_008772762.1 | XP_008770984.1 | 6 - 7 | 2390 | 131 | ||
GenBank mRNA | AF061971.1 | - | 6 - 7 | 698 | 131 | |
BC062023.1 | - | 6 - 7 | 728 | 131 | ||
FQ234132.1 | - | 6 - 7 | 708 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
