Assay Details
Gene Symbol: Ppt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256020.2 | XP_006256082.1 | 1 - 2 | 862 | 108 | |
GenBank mRNA | AF067790.1 | - | 1 - 2 | 109 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF067790.1, XM_006256020.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
