Assay Details
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165936.1 | NP_001159408.1 | 4 - 5 | 669 | 66 | |
NM_212532.2 | NP_997697.1 | 4 - 5 | 716 | 66 | ||
XM_008772714.1 | XP_008770936.1 | 5 - 6 | 815 | 66 | ||
XM_008772715.1 | XP_008770937.1 | 4 - 5 | 978 | 66 | ||
GenBank mRNA | BC086443.1 | - | 4 - 5 | 617 | 66 | |
BC097930.1 | - | 4 - 5 | 706 | 66 | ||
CO385374.1 | - | 4 - 5 | 716 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
