Assay Details
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165936.1 | NP_001159408.1 | 2 - 3 | 494 | 128 | |
NM_212532.2 | NP_997697.1 | 2 - 3 | 541 | 128 | ||
XM_008772714.1 | XP_008770936.1 | 3 - 4 | 640 | 128 | ||
XM_008772715.1 | XP_008770937.1 | 2 - 3 | 803 | 128 | ||
GenBank mRNA | BC086443.1 | - | 2 - 3 | 442 | 128 | |
BC097930.1 | - | 2 - 3 | 531 | 128 | ||
CO385374.1 | - | 2 - 3 | 541 | 128 | ||
EV770973.1 | - | 2 - 3 | 494 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
