Assay Details
Gene Symbol: Ddx39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133300.3 | NP_579834.2 | 9 - 10 | 1315 | 63 | |
GenBank mRNA | BC080243.1 | - | 9 - 10 | 1264 | 63 | |
FQ212901.1 | - | 9 - 10 | 1301 | 63 | ||
M75168.1 | - | 8 - 9 | 1220 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
