Assay Details
Gene Symbol: Egfl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004070.2 | NP_001004070.1 | 2 - 3 | 223 | 74 | |
NM_001165880.1 | NP_001159352.1 | 3 - 4 | 371 | 74 | ||
XM_006255977.2 | XP_006256039.1 | 3 - 4 | 1092 | 74 | ||
XM_006255978.2 | XP_006256040.1 | 4 - 5 | 978 | 74 | ||
XM_006255979.2 | XP_006256041.1 | 4 - 5 | 474 | 74 | ||
GenBank mRNA | BC127463.1 | - | 3 - 4 | 371 | 8362353 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
