Assay Details
Gene Symbol: Dxo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212497.2 | NP_997662.1 | 1 - 2 | 123 | 90 | |
GenBank mRNA | BC082083.1 | - | 1 - 2 | 123 | 90 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC082083.1, NM_212497.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
