Assay Details
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031021.3 | NP_112283.1 | 1 - 2 | 122 | 93 | |
GenBank mRNA | CB743000.1 | - | 1 - 2 | 142 | 93 | |
CB790784.1 | - | 1 - 2 | 168 | 93 | ||
EV778558.1 | - | 1 - 2 | 126 | 93 | ||
FQ214437.1 | - | 1 - 2 | 138 | 93 | ||
FQ224491.1 | - | 1 - 2 | 142 | 93 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_031021.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
