Assay Details
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035238.2 | NP_001030315.1 | 3 - 4 | 255 | 86 | |
NM_031021.3 | NP_112283.1 | 3 - 4 | 306 | 86 | ||
XM_006256081.2 | XP_006256143.1 | 3 - 4 | 342 | 86 | ||
GenBank mRNA | BC078807.1 | - | 3 - 4 | 255 | 86 | |
CB743000.1 | - | 3 - 4 | 326 | 86 | ||
CB790784.1 | - | 3 - 4 | 352 | 86 | ||
EV778558.1 | - | 3 - 4 | 310 | 86 | ||
FQ214437.1 | - | 3 - 4 | 322 | 86 | ||
FQ224491.1 | - | 3 - 4 | 326 | 86 | ||
FQ230013.1 | - | 3 - 4 | 294 | 86 | ||
HE864426.1 | - | 2 - 3 | 190 | 86 | ||
HE864431.1 | - | 190 | 86 | |||
HE864432.1 | - | 190 | 86 | |||
HE864433.1 | - | 190 | 86 | |||
HE864434.1 | - | 190 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
